Neßler-Reagenz, Tabletten
Lieferant: Lovibond
Synonyme: Potassium mercuric Iodide, Potassium tetraiodomercurate, Nota 1, Mercury (II) potassium iodide, Potassium tetraiodomercurate – Potassium hydroxide solution
511261BT 511231BT 511230BT 511251BT 511250BT
LOVI511261BTEA 123.8 EUR
LOVI511261BT LOVI511231BT LOVI511230BT LOVI511251BT LOVI511250BT
Neßler-Reagenz, Tabletten
Neßler-Reagenz
Molecular Weight: 786,4 g/mol
MDL: MFCD00049656
CAS-Nummer: 7783-33-7