Flashpoint tester, Abel
Lieferant: Stanhope Seta
Setaflash
STAN13032-3EA 0 EUR
STAN13032-3
Flashpoint tester, Abel
Flammpunktprüfer
The Abel flashpoint tester comprises of a copper water bath, an oil cup, a gas test jet, a manual stirrer and a timing pendulum.
Standards: BS 170, ISO 13736 (IP 74C and IP 75C thermometers included).
Lieferumfang: Supplied with thermometers for oil cup and water bath and instruction manual.