Canister filling stand
Lieferant: CEMO
CEMG10100EA 242 EUR
CEMG10100 CEMG8007
Canister filling stand
Stative
Filling stands, made from highly resistant stainless steel, suitable for acids and lyes.
  • Facilitates emptying of canisters up to 60 L
  • Cage secures mounting of canisters
  • Precise metering by swivelling of the canister