di-Natriumtetraborat, weiß, Pulver, Spectroflux® 200
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
12093.A1
12093.D9
12093.A1EA
0
EUR
12093.A1
12093.D9
di-Natriumtetraborat, weiß, Pulver, Spectroflux® 200
di-Natriumtetraborat
It can be used in testing of ceramics and steel.
Formel:
Na₂B₄O₇ Siedepunkt: 1575 °C Schmelzpunkt: 741 °C Dichte: 2,367 g/cm³ |
MDL:
MFCD00081185 CAS-Nummer: 1330-43-4 EINECS: 215-540-4 |
Spezifikation Testergebnisse
Appearance (Color) | White |
Form | Powder |
Na:B ratio | 1,009 - 1,116 |
Elemental Analysis - Boron, B (theoretical) | 21,49% |
Elemental Analysis - Sodium, Na (theoretical) | 22,85% |
Residue on Ignition | ≤1,0% (900 °C/30 minutes) |
Impurity Content, Silicon (Si) | ≤00 ppm |
Impurity Content, Others | ≤20 ppm max each |
Impurity Content, Total | ≤200 ppm |
Aluminium (Al) as Impurity | ≤50 ppm |
Calcium (Ca) as Impurity | ≤50 ppm |
Lithium (Li) as Impurity | ≤100 ppm |
Magnesium (Mg) as Impurity | ≤20 ppm |
Potassium (K) as Impurity | ≤50 ppm |
Sensitivity | Hygroscopic |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...