6-Maleinimido-hexansäure, Pulver

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: EMCA

46384.06 46384.14 46384.22
46384.06EA 57.7 EUR
46384.06 46384.14 46384.22
6-Maleinimido-hexansäure, Pulver
EMCA (6-Maleimidohexanoic acid)

6-Maleimidohexanoic acid is used as a probe for introducing maleimides groups into biomolecules and active pharmaceutical ingredients. It is also used with N-hydroxysuccinimide ester as a bifunctional cross-linking reagent. Acts as a probe for thiol groups in membrane proteins.


Soluble in water, methanol, ethanol, dimethyl sulfoxide. Insoluble in ether.


Incompatible with strong oxidizing agents, strong acids and strong bases and strong reducing agents

Formel: C₁₀H₁₃NO₄
Molecular Weight: 211,22 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00043140
CAS-Nummer: 55750-53-3

Bestellen

Spezifikation Testergebnisse

Form Powder
Solubility Soluble in water, methanol, ethanol, dimethyl sulfoxide. Insoluble in ether
Sensitivity Moisture Sensitive

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR