4-Phenyl-4-piperidinol 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Phenyl-4-piperidinol

H55418.03 H55418.06
H55418.03EA 59.1 EUR
H55418.03 H55418.06
4-Phenyl-4-piperidinol 99%
4-Phenyl-4-piperidinol
Formel: C₁₁H₁₅NO
Molecular Weight: 177,25 g/mol
Schmelzpunkt: 157…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006000
CAS-Nummer: 40807-61-2
EINECS: 255-089-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR