2-Keto-D-gluconic acid hemicalcium salt monohydrate 99%, Pulver
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Calcium 2-keto-D-gluconate, D-Arabino-2-hexulopyranosonic acid hemicalcium Salt
J66609.03
J66609.06
J66609.03EA
61.2
EUR
J66609.03
J66609.06
2-Keto-D-gluconic acid hemicalcium salt monohydrate 99%, Pulver
2-Keto-D-gluconic acid hemicalcium salt monohydrate
Formel:
C₆H₉O₇·½Ca·H₂O Molecular Weight: 231,22 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00149104 CAS-Nummer: 3470-37-9 |
Spezifikation Testergebnisse
Form | Powder |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...