D(+)-Trehalose Dihydrat 99, powder_white Cell Culture
Lieferant: MP Biomedicals
Synonyme: Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate
0219475605 0219475625 0219475680
ICNA0219475605EA 39.4 EUR
ICNA0219475605 ICNA0219475625 ICNA0219475680
D(+)-Trehalose Dihydrat 99, powder_white Cell Culture
D(+)-Trehalose Dihydrat
Trehalose Dihydrate is an osmolyte, a chemical chaperone. It contains many properties that allow it to stabilize partially unfolded protein molecules and inhibit protein aggregation. It include a low chemical reactivity, high glass transition temperature, existence of numerous polymorphs, nonreducing nature, and high affinity for water molecules. Additionally, Trehalose has been noted to bind to expanded polyglutamines and stabilize partially unfolded polyglutamine-containing proteins.
Trehalose, Dihydrate, is an inducer of autophagy, it protect cells against numerous environmental stresses. It is seldom used as a direct replacement for conventional sweeteners. Use as a cryoprotectant in a variety of cell freezing media.
Formel: C₁₂H₂₂O₁₁·2H₂O
Molecular Weight: 378.33 g/mol
Schmelzpunkt: 203 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00071594
CAS-Nummer: 6138-23-4
EINECS: 202-739-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af741ee934bfe7c82395d31c21e2c19f0738d0f0&vid=59183c854dcfa367e15d46861ef4b9a7028540a9&prid=web&clid=SAPDAM
Bestellen
Spezifikation Testergebnisse
Physical Appearance White Powder
Solubility Soluble in water (50 mg/mL - clear, colorless solution) or hot alcohol.