Sucralose 98%, Pulver

Lieferant: Thermo Scientific

Synonyme: Trichlorosucrose, (2R,3R,4R,5R,6R)-2-(((2R,3S,4S,5S)-2,5-Bis(chloromethyl)-3,4-dihydroxytetrahydrofuran-2-yl)oxy)-5-chloro-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4-diol, 1-(1,6-Dichloro-1,6-dideoxy-beta-D-fructofuranosyl)-4-chloro-4-deoxy-alpha-D-galactopyranoside

J66736.18 J66736.22 J66736.30
J66736.18EA 82.2 EUR
J66736.18 J66736.22 J66736.30
Sucralose 98%, Pulver
Sucralose

A synthetic sweetener approximately 600 times sweeter than sucrose. Activates T1R2/T1R3 sweet taste receptors on enteroendocrine cells

Formel: C₁₂H₁₉Cl₃O₈
Molecular Weight: 397,63 g/mol
Schmelzpunkt: 130 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD03648615
CAS-Nummer: 56038-13-2
EINECS: 259-952-2
Merck Index: 14,8880

Bestellen

Spezifikation Testergebnisse

Assay (unspecified) 97,5%
Optical Rotation +84 to +88°, c=1 in water

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR