D-Glucose-6-Phosphate Dipotassium Salt Hydrate
Lieferant: MP Biomedicals
0210031301
02100313.5
ICNA0210031301EA
142
EUR
ICNA0210031301
ICNA02100313.5
D-Glucose-6-Phosphate Dipotassium Salt Hydrate
D-Glucose-6-phosphate dipotassium salt hydrate
Storage temperature: -20 °C
Formel:
C₆H₁₁K₂O₉P |
CAS-Nummer:
5996-17-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...