Hydrocortison-21-acetat
Lieferant: MP Biomedicals
|
Achtung
|
Synonyme:
Hydrocortisone acetate
0210199505
0210199510
ICNA0210199505EA
137
EUR
ICNA0210199505
ICNA0210199510
Hydrocortison-21-acetat
Hydrocortison-21-acetat
HYDROCORTISONE-21-ACETATE (Δ4-Pregnen-11β,17α,21-triol- 3,20-dione; Cortisol acetate) Crystalline
Formel:
C₂₃H₃₂O₆ Molecular Weight: 404,51 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037714 CAS-Nummer: 50-03-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...