Probucol ≥98%

Lieferant: MP Biomedicals

Synonyme: 4,4'-(Isopropylidendithio)bis[2,6-di-tert-butylphenol], 4,4'-(Propane-2,2-diylbis(sulphanediyl))bis(2,6-di-tert-butylphenol)

0215637105 0215637110 0215637150
ICNA0215637105EA 139 EUR
ICNA0215637105 ICNA0215637110 ICNA0215637150
Probucol ≥98%
Probucol
Formel: C₃₁H₄₈O₂S₂
Molecular Weight: 516,84 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00079281
CAS-Nummer: 23288-49-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR