Erythrosin B Dinatriumsalz, rotes Pulver
Lieferant: MP Biomedicals
Achtung
Synonyme: Erythrosin 239C.I. 45430
ICNA0219044910EA 24.1 EUR
ICNA0219044910 ICNA0219044925 ICNA0219044905
Erythrosin B Dinatriumsalz, rotes Pulver
Erythrosin B Dinatriumsalz
A red, anionic xanthene dye, has been used as a contrast or counter stain to hematoxylin, crystal violet and other nuclear stains, in both plant and animal tissue.
Erythrosin B Sodium Salt is used as a counter stain to hematoxylin, crystal violet and other nuclear stains, in both plant and animal tissue. Erythosin B dye solution in 0.9% saline is made for intravenous injection to induce occlusion.
  • Concentration: 0.91
  • Soluble in water (20 mg/mL) or ethanol (20 mg/mL)
  • Room Temperature, desiccate
Formel: C₂₀H₆I₄Na₂O₅
Molecular Weight: 879,86 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00144257
CAS-Nummer: 16423-68-0
Bestellen
Spezifikation Testergebnisse
Identity Test Passes
UV/VIS: l max (water) 524-528nm