Clindamycin hydrochloride
Lieferant: MP Biomedicals
Synonyme: Cleocin Hydrochloride, (2S,4R)-N-((1S,2S)-2-Chloro-1-((2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(methylthio)tetrahydro-2H-pyran-2-yl)propyl)-1-methyl-4-propylpyrrolidine-2-carboxamide hydrochloride, 7[S]-Chloro-7-deoxylincomycin, Clindamycin HCl
0219116850 0219116810 0219116880
ICNA0219116850EA 394 EUR
ICNA0219116850 ICNA0219116810 ICNA0219116880
Clindamycin hydrochloride
Clindamycin hydrochloride
Synonyms: 7[S]-Chloro-7-deoxylincomycin; Cleocin Hydrochloride
Storage temperature.: +4 °C
Formel: C₁₈H₃₃ClN₂O₅S·HCl
Molecular Weight: 461.45 g/mol
MDL: MFCD07793327
CAS-Nummer: 21462-39-5