DNS Natriumsalz (Desoxyribonukleinsäure Natriumsalz) calf thymus

Lieferant: MP Biomedicals

Synonyme: Thymonucleic acid sodium salt, DNA sodium salt

0219512950 0219512990 0219512991
ICNA0219512950EA 47 EUR
ICNA0219512950 ICNA0219512990 ICNA0219512991
DNS Natriumsalz (Desoxyribonukleinsäure Natriumsalz) calf thymus
DNA sodium salt (Deoxyribonucleic acid sodium salt)

Polynucleotide, carrier of genetic material.


  • White Fibrous Solid
  • Soluble in water or aqueous buffers


The addition of SDS or similar detergent may help solubilize DNA; however, detergents tend to denature the DNA into shorter strands.


Calf thymus DNA (CT-DNA) is a natural DNA widely used in studies of DNA binding anticancer agents and DNA binding agents that modulate DNA structure and function. Calf thymus DNA is also used in physicochemical studies of DNA behavior in solution.


Synonyms: DNA sodium salt from calf thymus; Thymonucleic acid sodium salt; ctDNA

MDL: MFCD00130922
CAS-Nummer: 9007-49-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR