D(+)-Saccharose 99,9, ultrapure
Lieferant: MP Biomedicals
Synonyme: Haushaltszucker, D(+)-Sucrose, Rübenzucker, Rohrzucker, 2-β-D-Fructofuranosyl-1-α-D-glucopyranosid, Sucrose, Kristallzucker
04821713 04821721
ICNA04821713EA 126 EUR
ICNA04821713 ICNA04821721
D(+)-Saccharose 99,9, ultrapure
D(+)-Saccharose
Heavy metals (as Pb): <5 ppm
Glucose: <0.1%
Arsenic: <0.001 ppm
DNAse and RNAse-free.
Formel: C₁₂H₂₂O₁₁
Molecular Weight: 342.3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006626
CAS-Nummer: 57-50-1
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM