DL-sulphoraphane-Lösung ≥98%
Lieferant: Cayman Chemical
Gefahr
Synonyme: 1-Isothiocyanato-4-(methylsulphinyl)butane, 1-Isothiocyanato-4-methylsulphinyl)butane, sulphoraphane
10496-5 10496-10 10496-25
CAYM10496-5EA 55 EUR
CAYM10496-5 CAYM10496-10 CAYM10496-25
DL-sulphoraphane-Lösung ≥98%
DL-sulphoraphane
Sulforaphane is an isothiocyanate derived from cruciferous vegetables, including broccoli, that potently induces chemopreventative enzymes through Keap1-Nrf2 signaling and ARE-driven gene expression. At 15 μM, sulforaphane inhibits class I and II HDAC activity and suppresses tumor growth by inducing cell cycle arrest and apoptosis selectively in various cancerous prostate epithelial cells without affecting normal cells.
Formel: C₆H₁₁NOS₂
Molecular Weight: 177,3 g/mol
MDL: MFCD00198068
CAS-Nummer: 4478-93-7
UN: 1170
ADR: 3,II