Adapalene ≥98%

Lieferant: Cayman Chemical

Synonyme: 6-[3-(1-Adamantyl)-4-methoxyphenyl]-2-naphthoic Acid, 6-(3-(Adamantan-1-yl)-4-methoxyphenyl)-2-naphthoic acid

13655-100 13655-250 13655-500 13655-1
CAYM13655-100EA 79 EUR
CAYM13655-100 CAYM13655-250 CAYM13655-500 CAYM13655-1
Adapalene ≥98%
Adapalene

Adapalene, a synthetic naphthoic acid derivative, is a selective agonist of retinoic acid receptor (RAR)-β and RAR-γ (Kds = 34 and 130 nM, respectively). At 0,1-10 µM, adapalene dose-dependently inhibits sebocyte growth, proliferation and differentiation in a primary rat preputial cell culture system. Adapalene demonstrates anti-inflammatory and immunomodulatory activity by inhibiting 5-lipoxygenase-catalyzed leukotriene production and expression of toll-like receptor 2 on human monocytes. It is commonly used for the study of the pathogenesis and treatment of acne vulgaris.

Formel: C₂₈H₂₈O₃
Molecular Weight: 412,53 g/mol
MDL: MFCD03106112
CAS-Nummer: 106685-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR