Glasröhren
Lieferant: Schott AG
SCOR1485590EA 0 EUR
SCOR1485590 SCOR1518580 SCOR1485595 SCOR1194020 SCOR1391244 SCOR1391200 SCOR1504772 SCOR1564520 SCOR1518606 SCOR1028943 SCOR1487826 SCOR1028081 SCOR1194016 SCOR1504767 SCOR1028162 SCOR1545315 SCOR1193988 SCOR1193964 SCOR1193986 SCOR1194018 SCOR1193985 SCOR1027897 SCOR1193963 SCOR1193962 SCOR1194017 SCOR1263533 SCOR1545822 SCOR1579276 SCOR1270387 SCOR1552451 SCOR1404444 SCOR1402201 SCOR1552454
Glasröhren
Schläuche Transferschlauch
AR-Glas®, clear.
  • Good chemical and physical properties
  • Suitable for the production of items that must normally withstand only brief exposure to aggressive chemicals
  • Not resistant to high thermal loads
Tube length ~1500 mm.