cis-3-Hexenal-Lösung 50% in Triacetin stabilisiert für die Nahrungsmittelindustrie Halal, Kosher, Sigma-Aldrich®

Lieferant: Merck


Stabilisierung: Stabilisiert mit α-Tocopherol
M-Clarity™ Qualitätsstufe = MQ300

W256102-100G-K W256102-1KG-K W256102-25G-K
SIALW256102-100G-KEA 1750 EUR
SIALW256102-100G-K SIALW256102-1KG-K SIALW256102-25G-K
cis-3-Hexenal-Lösung 50% in Triacetin stabilisiert für die Nahrungsmittelindustrie Halal, Kosher, Sigma-Aldrich®
cis-3-Hexenal

It contains synthetic α-tocopherol as stabiliser.


cis-3-Hexenal solution may be used as a standard to identify and quantify the volatile organic compounds emitted by wounded plants using chemical-ionization mass spectrometry

Formel: C₆H₁₀O
Molecular Weight: 98,14 g/mol
Siedepunkt: 20 °C (0,2 mmHg)
Dichte: 0,982 g/ml (25 °C)
Lagertemperatur: Gefrierschrank
CAS-Nummer: 6789-80-6
UN: 1207
ADR: 3,III

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Colourless to Faint Yellow
Form Liquid
Refractive index at 20 °C 1,430 - 1,442
Infrared spectrum Conforms to Structure
Titration by hydroxylamine 45,0 - 55,0%
Color test (APHA) 300 APHA maximum
Arsenic (As) ≤3 ppm
Cadmium (Cd) ≤1 ppm
Mercury (Hg) ≤1 ppm
Lead (Pb) ≤10 ppm
Concentration 50% in triacetin
GMO content GMO free

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR