Anti-Borrelia burgdorferi Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BOSSBS-12879R-A350EA 842 EUR
BOSSBS-12879R-A350
Anti-Borrelia burgdorferi Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Borrelia burgdorferi is a spirochete and the cause of Lyme disease, a tick transmitted illness of humans and animals. B. burgdorferi may persist in humans and animals for months or years following initial infection, despite a robust humoral immune response. B. burgdorferi resembles other spirochetes in that it is a highly specialized, motile, two-membrane, spiral shaped bacteria which lives primarily as an extracellular pathogen. B. burgdorferi has an unusual genome compared with other eubacteria which includes a linear chromosome approximately one megabase in size and numerous linear and circular plasmids.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Borrelia burgdorferi
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR