Anti-Mycobacterium bovis Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BOSSBS-4622R-A555EA 842 EUR
BOSSBS-4622R-A555
Anti-Mycobacterium bovis Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Mycobacterium bovis

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Mycobacterium bovis
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR