Anti-S. aureus and R. tropina Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BOSSBS-4582R-HRPEA 858 EUR
BOSSBS-4582R-HRP
Anti-S. aureus and R. tropina Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Staphyloccocus aureus Rosenbach tropina

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: Staphyloccocus aureus Rosenbach tropina
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR