Levamisole hydrochloride ≥99% (durch GC), weißes Pulver, Sigma-Aldrich®
Lieferant: Merck
Gefahr
Synonyme: (-)-Tetramisole Hydrochloride, (-)-2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole Hydrochloride, (S)-6-Phenyl-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole hydrochloride
M-Clarity™ Qualitätsstufe = MQ200
L9756-10G L9756-5G
SIALL9756-10GEA 145 EUR
SIALL9756-10G SIALL9756-5G
Levamisole hydrochloride ≥99% (durch GC), weißes Pulver, Sigma-Aldrich®
Levamisole hydrochloride
Suitable for inhibition of various mammalian alkaline phosphatase (i.e., liver, kidney, placenta, bone and tumor).
  • Intestinal alkaline phosphatases are only slightly inhibited
  • Not for veterinary use
Formel: C₁₁H₁₂N₂S·HCl
Molecular Weight: 240.75 g/mol
Schmelzpunkt: 228…230 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00012675
CAS-Nummer: 16595-80-5
UN: 2811
ADR: 6.1 III