(S)-(+)-Camptothecin ≥90% (durch HPLC), Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, (+)-Camptothecin, 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione

M-Clarity™ Qualitätsstufe = MQ200

C9911-100MG C9911-1G C9911-250MG
SIALC9911-100MGEA 302 EUR
SIALC9911-100MG SIALC9911-1G SIALC9911-250MG
(S)-(+)-Camptothecin ≥90% (durch HPLC), Sigma-Aldrich®
(S)-(+)-Camptothecin

Camptothecin is an alkaloid that exhibits anti-leukemic and anti-tumor activities.


  • Featured product for apoptosis research

Formel: C₂₀H₁₆N₂O₄
Molecular Weight: 348.36 g/mol
Schmelzpunkt: 260 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00081076
CAS-Nummer: 7689-03-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR