7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene ≥98% (durch GC), Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
MTBD, 1,3,4,6,7,8-Hexahydro-1-methyl-2H-pyrimido[1,2-a]pyrimidine
M-Clarity™ Qualitätsstufe = MQ200
7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene is a soluble amine base. It forms 1:1 complexes with 4-nitrophenyl[bis(diethylsulfonyl)]methane and phenyl[bis(diethylsulfonyl)]methane.
Used in microwave-promoted, palladium-catalysed C-N bond-forming reactions with aryl/heteroaryl nonaflates and amines also used as strong base to investigate the podand solvents, tris(oxaalkyl)phenylsilanes and tris(oxaalkyl)phosphates.
Synonyms: 1,3,4,6,7,8-Hexahydro-1-methyl-2H-pyrimido[1,2-a]pyrimidine, MTBD.
Formel:
C₈H₁₅N₃ Molecular Weight: 153,23 g/mol Siedepunkt: 75,..79 °C (0.1 mmHg)) (Lit.) Dichte: 1,067 g/ml (25 °C) (Lit.) |
MDL:
MFCD00043004 CAS-Nummer: 84030-20-6 UN: 3267 ADR: 8,II |
Spezifikation Testergebnisse
Assay | 98% |
Refractive index | n20/D 1,537 (lit.) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...