3,4-Dimethyl-1H-pyrazole 97%, kristallines Pulver
Lieferant: Thermo Scientific
|
Gefahr
|
H37099.14
H37099.06
H37099.14EA
361
EUR
H37099.14
H37099.06
3,4-Dimethyl-1H-pyrazole 97%, kristallines Pulver
3,4-Dimethyl-1H-pyrazole
Used as pharmaceutical intermediate.
- Incompatible with oxidising agents
- Synonym: 3,4-Dimethylpyrazole
Formel:
C₅H₈N₂ Molecular Weight: 96,133 g/mol Schmelzpunkt: 53…55 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00462154 CAS-Nummer: 2820-37-3 EINECS: 429-130-1 |
Spezifikation Testergebnisse
Appearance (Colour) | White to cream |
Form | Crystalline powder |
Assay (GC) | >96,0% |
Solubility | Slightly soluble in water |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...