Ubiquinone-5-Lösung ≥98%
Lieferant: Cayman Chemical
Synonyme: Coenzyme Q1
18741-1 18741-10 18741-5
CAYM18741-1EA 65 EUR
CAYM18741-1 CAYM18741-10 CAYM18741-5
Ubiquinone-5-Lösung ≥98%
Ubiquinone-5
This Ubiquinone including CoQ1, impact mitochondrial permeability transition pore (PTP) formation, as well as PTP-dependent cell death, in an analog- and cell-specific manner.
  • Solution in ethanol
CoQ1 is an amphipathic CoQ10 homolog that has a tail consisting of five isoprene units. It has been used as an electron acceptor to study a range of oxidoreductases as isolated enzymes, in subcellular fractions, in intact cells in culture, and in perfused organs.
Formula: 2,3-dimethoxy-5-methyl-6-(3-methyl-2-buten-1-yl)-2,5-cyclohexadiene-1,4-dione
Formel: C₁₄H₁₈O₄
Molecular Weight: 250.3 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 727-81-1