(2,4-Dichlorophenoxy)acetic acid sodium salt monohydrate ≥98% (durch GC), BioReagent Geeignet für Pflanzenzellkulturen, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: 2,4-D sodium salt monohydrate

M-Clarity™ Qualitätsstufe = MQ200

D6679-25G D6679-5G
SIALD6679-25GEA 534 EUR
SIALD6679-25G SIALD6679-5G
(2,4-Dichlorophenoxy)acetic acid sodium salt monohydrate ≥98% (durch GC), BioReagent Geeignet für Pflanzenzellkulturen, Sigma-Aldrich®
(2,4-Dichlorophenoxy)acetic acid sodium salt monohydrate

2,4-Dichlorophenoxyacetic acid (2,4-D) is a synthetic auxin (plant hormone) used in plant cell culture media such as Murashige and Skoog media. 2,4-D is also a widely used herbicide.

Formel: C₈H₅Cl₂NaO₃·H₂O
Molecular Weight: 261,03 g/mol
MDL: MFCD01311802
CAS-Nummer: 7084-86-8
UN: 3077
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Appearance (Color) White to Off-White
Appearance (Form) Powder
Solubility (Color) Colorless
Solubility (Turbidity, 50 mg/ml, H₂O) Clear
Water (by Karl Fischer) ≤10 %
Carbon (anhydrous) 37.5 - 41.3 %
Sodium (Na) anhydrous 7.1 - 11.8 %
Purity (GC) ≥98 %
Plant Cell Culture Test Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR