Ethylacetat, AR®, Laboratory Reagent, Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
Synonyme: Essigester, Essigsäureethylester, EtOAc
6780-45EA 424.79 EUR
6780-45 6780-25
Ethylacetat, AR®, Laboratory Reagent, Macron Fine Chemicals™
Ethylacetat
Formel: CH₃COOC₂H₅
Molecular Weight: 88.11 g/mol
Siedepunkt: 77.1
Schmelzpunkt: -83
Flammpunkt: -4
MDL: MFCD00009171
CAS-Nummer: 141-78-6
UN: 1173
ADR: 3 II
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM