Kaliumcarbonat, Granulat FCC, J.T.Baker®

Lieferant: Avantor

Synonyme: Pottasche

3014-05EA 377 EUR
3014-05 3014-08
Kaliumcarbonat, Granulat FCC, J.T.Baker®
Kaliumcarbonat
Formel: K₂CO₃
MDL: MFCD00011382
CAS-Nummer: 584-08-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Assay (K₂CO₃) (dried basis) 99.0 - 100.5 %
FCC - Loss on Drying ≤1 %
FCC - Insoluble Substances Passes Test
FCC - Lead (Pb) ≤2 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR