Kaliumpermanganat 0.0995-0.1005 N in wässriger Lösung, BAKER ANALYZED®, Laborreagens volumetrische Lösung, J.T.Baker®
Lieferant: Avantor
Synonyme: Permanganic acid potassium salt
5651-02EA 0 EUR
5651-02 5651-03
Kaliumpermanganat 0.0995-0.1005 N in wässriger Lösung, BAKER ANALYZED®, Laborreagens volumetrische Lösung, J.T.Baker®
Potassium permanganate
Standardised at 25 °C against chemicals whose certification is traceable to NIST. Lot standardisation value, accurate to ±1 part per 1000, provided on label.
Formel: KMnO₄
Molecular Weight: 158,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011364
CAS-Nummer: 7722-64-7
EINECS: 231-760-3
UN: 3082
ADR: 9,III
Bestellen
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use
Normality 0.0995 - 0.1005
Chloride and Chlorate (as Cl) ≤10 ppm
Sulfate (SO₄) ≤20 ppm
Standardization at 25°C traceable to NIST Standard Reference Material.
SRM No…