Piroxicam ≥95%

Lieferant: Cayman Chemical

Synonyme: 4-Hydroxy-2-methyl-N-(2-pyridyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, 4-Hydroxy-2-methyl-N-(pyridin-2-yl)-2H-benzo[e][1,2]thiazine-3-carboxamide 1,1-dioxide, 4-Hydroxy-2-methyl-3-(2-pyridylcarbamoyl)-2H-1,2-benzothiazine 1,1-Dioxide

13368-500 13368-1 13368-5 13368-10
CAYM13368-500EA 27 EUR
CAYM13368-500 CAYM13368-1 CAYM13368-5 CAYM13368-10
Piroxicam ≥95%
Piroxicam

Piroxicam is a non-steroidal anti-inflammatory drug which inhibits both isoforms of cyclooxygenase in a non selectable and reversible manner. IC50 = 1,57 and 1,69 μM for COX-1 and COX-2, respectively.


It has an extended half-life of about 40 hours, which allows once daily administration leading to sustained plasma and tissue levels. In addition to its anti-inflammatory effects, piroxicam is known to be an effective analgesic that has been used extensively in the treatment of arthritis. It has also been used to inhibit COX activity for treatment of certain cancers in animals. As with other NSAIDs, the extended use of piroxicam results in damage to the gastrointestinal lining.

Formel: C₁₅H₁₃N₃O₄S
Molecular Weight: 331,35 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00057317
CAS-Nummer: 36322-90-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR