4-Methylumbelliferyl-β-D-xylopyranoside ≥98% (durch HPLC), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Methyl-7-(((2S,3R,4S,5R)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one, 4-Methylumbelliferyl-β-D-xylopyranoside, β-xylosidase substrate

M-Clarity™ Qualitätsstufe = MQ200

M7008-100MG M7008-1G M7008-250MG M7008-25MG
SIALM7008-100MGEA 251 EUR
SIALM7008-100MG SIALM7008-1G SIALM7008-250MG SIALM7008-25MG
4-Methylumbelliferyl-β-D-xylopyranoside ≥98% (durch HPLC), Sigma-Aldrich®
4-Methylumbelliferyl-beta-D-xylopyranoside

A substrate for β-xylosidase.

Formel: C₁₅H₁₆O₇
Molecular Weight: 308,286 g/mol
MDL: MFCD00037607
CAS-Nummer: 6734-33-4

Bestellen

Spezifikation Testergebnisse

Appearance (color) White
Appearance (form) Powder
Solubility (color) Colorless to faint yellow
Solubility, 50 mg/ml in Pyridine (turbidity) Clear
Infrared spectrum Conforms to structure
Carbon 57,3 - 59,6%
Purity (HPLC) ≥98%
Free methylumbelliferone (impurity) ≤0,10%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR