Paroxetine Related Compounds, Reference Standard

Lieferant: USP

1500218 1500230 1500240 1500251 1500262 1500273 1500284 1500353 1500207
1500218.EA 385 EUR
1500218. 1500230. 1500240. 1500251. 1500262. 1500273. 1500284. 1500353. 1500207.
Paroxetine Related Compounds, Reference Standard
Standards Organische Standards

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.


Standards: US Pharmacopea Reference Standard

Bestellen

TECHNISCHE DATEN

Bezeichnung Paroxetine Hydrochloride Paroxetine Related Compound B Paroxetine Related Compound C Paroxetine Related Compound D Paroxetine Related Compound F Paroxetine Related Compound G Paroxetine A Paroxetine Related Compound E
Environmentally Preferable
Synonym(s) (3S,4R)-3-[(Benzodioxol-5-yloxy)methyl]-4-phenylpiperidine hydrochloride (3R,4S)-3-[(Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine hydrochloride) OR ((+)-trans-Paroxetine hydrochloride (3RS,4RS)-3-[(Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine hydrochloride); cis-paroxetine hydrochlorid (3S,4R)-3-[(Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-1-methylpiperidine (+/-)trans-3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4''-fluorophenyl-4'-phenyl)piperidine hydrochloride 4-(4-Fluorophenyl)-1-methyl-1,2,3,6-tetrahydropyridine hydrochloride
CAS Nr. 110429-35-1 130855-30-0 110429-36-2 1012886-75-7
Formel C₃₈H₄₄Cl₂F₂N₂O₇ C₁₉H₂₁ClFNO₃ C₂₀H₂₂FNO₃
UN No. 3077 3077

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR