(+)-Limonene 1,2-epoxide cis- und trans-Gemisch ≥97,0% (Summe der Isomere), Sigma-Aldrich®
Lieferant: Merck
Synonyme: (4R)-1,2-Epoxy-p-menth-8-ene
17763-100ML 17763-500ML
SIAL17763-100MLEA 160 EUR
SIAL17763-100ML SIAL17763-500ML
(+)-Limonene 1,2-epoxide cis- und trans-Gemisch ≥97,0% (Summe der Isomere), Sigma-Aldrich®
(+)-Limonene 1,2-epoxide
(+)-Limonene 1,2-epoxide is a monoterpene that can be prepared from (+)-limonene has mixture of cis/trans-isomers. It shows potent antitumoral and antinociceptive activity. The main application is used as building block in synthesis of pharmaceutical products, perfumes, cosmetics and food additives.
  • Density: 0,930 g/ml at 20°C (lit.)
  • Refractive index: n20/D 1,467
  • Structure is confirmed by proton NMR spectrum
  • colorless to very faint yellow and colorless to very faint green-yellow, liquid
Molecular Weight: 152,23 g/mol
CAS-Nummer: 203719-54-4