Kaliumhydrogenphthalat Standardlösung 0,2125% (w/v) in Wasser

Lieferant: Bernd Kraft

01282.3000 01282.2000
KRAF01282.3000EA 93.2 EUR
KRAF01282.3000 KRAF01282.2000
Kaliumhydrogenphthalat Standardlösung 0,2125% (w/v) in Wasser
Kaliumhydrogenphthalat

2,125 g C8H5KO4 in 1 l of water for determination of TOC and DOC according to EN 1484:1997, DEV H3.

Formel: HOOCC₆H₄COOK
MDL: MFCD00013070
CAS-Nummer: 877-24-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4652479e822d1591ac7eb5e29f892b8c94588ad4&vid=75071da7f30b07f7bd077699a3b54ec89d80f0b8&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR