Organische Referenzstandards, Candesartan cilexetile, RS
Lieferant: USP
1087803 1087869
1087803.EA 1160 EUR
1087803. 1087869.
Organische Referenzstandards, Candesartan cilexetile, RS
Standards Organische Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Standards: US Pharmacopea Reference Standard
Bestellen
TECHNISCHE DATEN
Bezeichnung Candesartancilexetil Candesartan Cilexetil
Synonym(s) 1-(Cyclohexyloxycarbonyloxy)ethyl 2-ethoxy-1-{[2'-(2-ethyl-2H-tetrazol-5-yl)biphenyl-4-yl]methyl}-1H-benzimidazole-7-carboxylate
Environmentally Preferable
CAS Nr. 145040-37-5 914613-36-8
Formel C₃₃H₃₄N₆O₆ C₃₅H₃₈N₆O₆