Halbhohe ESD-Sicherheitsschuhe, zum Schnüren, New High
Lieferant: MILLE S.A.S
NHHG3/45 NHHG3/39 NHHG3/40 NHHG3/41 NHHG3/42 NHHG3/44 NHHG3/46 NHHG3/47 NHHG3/43
MILENHHG3/45EA 113 EUR
MILENHHG3/45 MILENHHG3/39 MILENHHG3/40 MILENHHG3/41 MILENHHG3/42 MILENHHG3/44 MILENHHG3/46 MILENHHG3/47 MILENHHG3/43
Halbhohe ESD-Sicherheitsschuhe, zum Schnüren, New High
Schuhe und Stiefel
These anti-static and metal-free safety shoes are made from a very sturdy, water and oil resistant suede leather upper (2-2, 2 mm) and an extra light PU dual-density outer sole.
  • Abrasion-resistant, antibacterial poromax® lining with high water-absorbing capacity for additional breathability
  • Waterproof tongue and heel grip
  • Composite toe protection cap (240 J)
  • Textile puncture resistant midsole (1 430 N)
Colour: Grey
Suitable for industries, finishing construction, logistics and services.
Standards: EN ISO 20345; IEC EN 61340-5-1 (ESD); S3, SRC