Anti-LAG3 Mouse Monoclonal Antibody (ATTO 647N) [clone: 17B4]

Lieferant: ProSci
PRSI36-113EA 612 EUR
PRSI36-113
Anti-LAG3 Mouse Monoclonal Antibody (ATTO 647N) [clone: 17B4]
Antikörper
Lymphocyte activation gene 3 (LAG-3; CD223) plays an important role in negatively regulating T cell proliferation, function and homeostasis. It is required for maximal natural and induced regulatory T cell (Treg) function. LAG-3 is closely related to the T cell co-receptor CD4 and binds to MHC class II molecules but with a significantly higher affinity than CD4.

Functional Application: Inhibition. Immunohistochemistry: (frozen sections (1:150)). Immunoprecipitation: (10ug/ml). Western Blot: (5ug/ml).

Type: Primary
Antigen: LAG3
Clonality: Monoclonal
Clone: 17B4
Conjugation: ATTO 647N
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR