Filter cartridges, Sartoguard® GF
Lieferant: Sartorius
5488307G2-SS 5485307G9-SS-A 5485307G8-SS-A 5485307G0-SS-V 5488307G1-SS 5488307G3-SS
SARP5488307G2-SSEA 785 EUR
SARP5488307G2-SS SARP5485307G9-SS-A SARP5485307G8-SS-A SARP5485307G0-SS-V SARP5488307G1-SS SARP5488307G3-SS
Filter cartridges, Sartoguard® GF
Filter Filterkartuschen
Sartoguard® GF filter elements consist of glass fiber layers and PES membranes. They combine the defined retention performance for particles and microorganisms of membrane filters with the high total throughput capacity in the depth of the fibrous material. They are ideally suited for effective retention of particles, colloids and lipids which block more expensive sterilising grade or Mycoplasma retentive membrane filters quickly.
  • High adsorptive power due to glass fiber fleeces
  • Protection of downstream equipment and final membrane filters
  • Bioburden
More Product Information