Salicylaldehydazin 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: Salicylaldazine

456940050 456940250 456941000
ACRO456940050EA 52.1 EUR
ACRO456940050 ACRO456940250 ACRO456941000
Salicylaldehydazin 99%
Salicylaldehydazin
Formel: C₁₄H₁₂N₂O₂
Molecular Weight: 240,26 g/mol
Schmelzpunkt: 218…220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00043496
CAS-Nummer: 959-36-4

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to brown Powder
Infrared spectrum Conforms
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR