Uridine-5'-diphosphoglucuronic acid trisodium salt 98-100%, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Sodium (2S,3S,4S,5R,6R)-6-(((((((2R,3S,4R,5R)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)oxidophosphoryl)oxy)oxidophosphoryl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylate, UDP-glucuronic acid trisodium, UDP-GlcA, Tripotassium 6-[({[5-(2,4-dioxo-3H-pyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxyphosphinato)oxy]-3,4,5-trihydroxyoxane-2-carboxylate, UDP-D-GLUCURONIDE TRISODIUM SALT, UDPGA

U6751-100MG U6751-250MG U6751-25MG U6751-500MG U6751-1G
SIALU6751-100MGEA 380 EUR
SIALU6751-100MG SIALU6751-250MG SIALU6751-25MG SIALU6751-500MG SIALU6751-1G
Uridine-5'-diphosphoglucuronic acid trisodium salt 98-100%, Sigma-Aldrich®
Uridine-5'-diphosphoglucuronic acid trisodium salt
Formel: C₁₅H₁₉N₂Na₃O₁₈P₂
Molecular Weight: 646.23 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00037687
CAS-Nummer: 63700-19-6

Bestellen

Spezifikation Testergebnisse

Appearance (colour) White
Appearance (form) Powder
Solubility (colour) Colourless
Solubility (turbidity) 50 mg/ ml, H2O Clear
Phosphorus (dry basis) 9,0 - 10,1 %
Sodium (dry basis) 10,1 - 11,2 %
Water (by Karl Fischer) ≤10%
1H NMR spectrum Conforms to structure
Solvent content (by GC) ≤5%
Purity (HPLC) 98 - 100 %
Storage temperature –150 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR