Solvent recycler, cart-mounted
Lieferant: CBG Biotech
KPXXSLV-99-CE-20EA 30560 EUR
KPXXSLV-99-CE-20 KPXXSLV-99-CE-40
Solvent recycler, cart-mounted
Destillationsbauteile Destillations-Bausätze
These solvent recycler recycles xylene, xylene substitutes, alcohol and acetone with 90 - 95% recovery.
  • Fractional distillation
  • Automatic waste disposal
  • Microprocessor controlled, fully automated
  • Built-in spill containment
  • Mobile apparatus with electric fill pump
It does not require water hook-up or special ventilation and satisfies the latest uniform fire code requirements for solvent distillation units.
Cycle time: 15 to 20 min/l (Xylene) and 20 to 30 min/l (Ethanol)