Methyl-5-brom-2-furoat 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-Bromo-2-furoic acid methyll ester, 5-Bromo-2-furancarboxylic acid methyll ester, Methyl 5-Bromo-2-furancarboxylate, Methyl 5-Bromo-2-furoate, Methyl 5-bromofuran-2-carboxylate

355750010 355750050 355750250
ACRO355750010EA 33.7 EUR
ACRO355750010 ACRO355750050 ACRO355750250
Methyl-5-brom-2-furoat 98%
Methyl-5-brom-2-furoat
Formel: C₆H₅BrO₃
Molecular Weight: 205.01 g/mol
Schmelzpunkt: 65…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00092313
CAS-Nummer: 2527-99-3

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 63 to 68 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR