5-Fluorobenzoxazole 97%

Lieferant: Thermo Scientific
Achtung

455610010 455612500
ACRO455610010EA 269 EUR
ACRO455610010 ACRO455612500
5-Fluorobenzoxazole 97%
5-Fluorobenzoxazole
Formel: C7H4FNO
Molecular Weight: 137,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08458151
CAS-Nummer: 221347-71-3

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to brown Crystalline powder or needles
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR