6-Bromisatin 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Bromo-1H-indole-2,3-dione, 6-Bromoindoline-2,3-dione
455510050
455510250
ACRO455510050EA
67.7
EUR
ACRO455510050
ACRO455510250
6-Bromisatin 95%
6-Bromisatin
Formel:
C₈H₄BrNO₂ Molecular Weight: 226,03 g/mol Schmelzpunkt: 274 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01631138 CAS-Nummer: 6326-79-0 |
Spezifikation Testergebnisse
Appearance | Yellow to orange Powder |
Infrared spectrum | Conforms |
Melting point | 267 to 275 °C |
Loss on drying | ≤3 % |
HPLC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...