2-Brompyrazin 95%

Lieferant: Thermo Fisher Scientific
Gefahr

458160010 458160050
ACRO458160010EA 84.4 EUR
ACRO458160010 ACRO458160050
2-Brompyrazin 95%
2-Bromopyrazine
Formel: C₄H₃BrN₂
Molecular Weight: 158.9800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08275680
CAS-Nummer: 56423-63-3

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR