3,4-Dihydroisoquinolin-1(2H)-one 98%

Lieferant: Thermo Fisher Scientific

458220010 458220050
ACRO458220010EA 94.9 EUR
ACRO458220010 ACRO458220050
3,4-Dihydroisoquinolin-1(2H)-one 98%
3,4-Dihydroisoquinolin-1(2H)-one
Formel: C9H9NO
Molecular Weight: 147.1700 g/mol
Schmelzpunkt: 67…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00853963
CAS-Nummer: 1196-38-9

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR