Omeprazole 98+%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Methoxy-2-(((4-methoxy-3,5-dimethylpyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole
458520010
458520050
458520250
ACRO458520010EA
36.9
EUR
ACRO458520010
ACRO458520050
ACRO458520250
Omeprazole 98+%
Omeprazole
Formel:
C₁₇H₁₉N₃O₃S Molecular Weight: 345,42 g/mol Schmelzpunkt: 158…159 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00083192 CAS-Nummer: 73590-58-6 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % (60°C, 4 h) (vacuum) |
HPLC | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...